Anti-KLK8 Mouse Monoclonal Antibody [clone: 189004]

Supplier: R & D Systems
MAB1719
220019-291EA 763.57 USD
220019-291
Anti-KLK8 Mouse Monoclonal Antibody [clone: 189004]
Antibodies
Type: Primary
Antigen: KLK8
Clonality: Monoclonal
Clone: 189004
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR