Anti-CLEC1B Mouse Monoclonal Antibody [clone: 219150]

Supplier: R & D Systems
MAB1718
220019-290EA 763.57 USD
220019-290
Anti-CLEC1B Mouse Monoclonal Antibody [clone: 219150]
Antibodies
Type: Primary
Antigen: CLEC1B
Clonality: Monoclonal
Clone: 219150
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR