Anti-ADAMTSL1 Mouse Monoclonal Antibody [clone: 212125]

Supplier: R & D Systems
MAB1717
220019-289EA 763.57 USD
220019-289
Anti-ADAMTSL1 Mouse Monoclonal Antibody [clone: 212125]
Antibodies
Type: Primary
Antigen: ADAMTSL1
Clonality: Monoclonal
Clone: 212125
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR