Anti-IL23A Mouse Monoclonal Antibody [clone: 727753]

Supplier: R & D Systems
MAB17161
102885-294EA 600.15 USD
102885-294
Anti-IL23A Mouse Monoclonal Antibody [clone: 727753]
Antibodies
Type: Primary
Antigen: IL23A
Clonality: Monoclonal
Clone: 727753
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR