Anti-CNTN2 Mouse Monoclonal Antibody [clone: 372913]

Supplier: R & D Systems
MAB17141
220019-287EA 594.37 USD
220019-287
Anti-CNTN2 Mouse Monoclonal Antibody [clone: 372913]
Antibodies
Type: Primary
Antigen: CNTN2
Clonality: Monoclonal
Clone: 372913
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR