Anti-CSTA1 Rat Monoclonal Antibody [clone: 263423]

Supplier: R & D Systems
MAB1710
220019-285EA 763.57 USD
220019-285
Anti-CSTA1 Rat Monoclonal Antibody [clone: 263423]
Antibodies
Type: Primary
Antigen: CSTA1
Clonality: Monoclonal
Clone: 263423
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR