Anti-CXCR4 Mouse Monoclonal Antibody [clone: 44708]

Supplier: R & D Systems
MAB171-100
102885-292EA 633.42 USD
102885-292
Anti-CXCR4 Mouse Monoclonal Antibody [clone: 44708]
Antibodies
Type: Primary
Antigen: CXCR4
Clonality: Monoclonal
Clone: 44708
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR