Anti-LCN1 Mouse Monoclonal Antibody [clone: 223908]

Supplier: R & D Systems
MAB1708
220019-283EA 763.57 USD
220019-283
Anti-LCN1 Mouse Monoclonal Antibody [clone: 223908]
Antibodies
Type: Primary
Antigen: LCN1
Clonality: Monoclonal
Clone: 223908
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR