Anti-SIGLEC5 Rat Monoclonal Antibody [clone: 238023]

Supplier: R & D Systems
MAB1706
220019-279EA 763.57 USD
220019-279
Anti-SIGLEC5 Rat Monoclonal Antibody [clone: 238023]
Antibodies
Type: Primary
Antigen: SIGLEC5
Clonality: Monoclonal
Clone: 238023
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR