Anti-CLEC1A Mouse Monoclonal Antibody [clone: 219003]

Supplier: R & D Systems
MAB1704
220019-277EA 763.57 USD
220019-277
Anti-CLEC1A Mouse Monoclonal Antibody [clone: 219003]
Antibodies
Type: Primary
Antigen: CLEC1A
Clonality: Monoclonal
Clone: 219003
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR