Anti-GATA6 Mouse Monoclonal Antibody [clone: 222228]

Supplier: R & D Systems
MAB1700
220019-276EA 594.37 USD
220019-276
Anti-GATA6 Mouse Monoclonal Antibody [clone: 222228]
Antibodies
Type: Primary
Antigen Symbol: GATA6
Antigen Name: GATA binding protein 6
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 222228
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR