Anti-CXCR4 Mouse Monoclonal Antibody [clone: 12G5]

Supplier: R & D Systems
MAB170
220019-275EA 728.86 USD
220019-275
Anti-CXCR4 Mouse Monoclonal Antibody [clone: 12G5]
Antibodies
Type: Primary
Antigen: CXCR4
Clonality: Monoclonal
Clone: 12G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR