Anti-ITGAM Mouse Monoclonal Antibody [clone: 238439]

Supplier: R & D Systems
MAB16992
220019-274EA 772.25 USD
220019-274
Anti-ITGAM Mouse Monoclonal Antibody [clone: 238439]
Antibodies
Type: Primary
Antigen: ITGAM
Clonality: Monoclonal
Clone: 238439
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR