Anti-ITGAM Mouse Monoclonal Antibody [clone: 238446]

Supplier: R & D Systems
MAB16991
220019-273EA 594.37 USD
220019-273
Anti-ITGAM Mouse Monoclonal Antibody [clone: 238446]
Antibodies
Type: Primary
Antigen: ITGAM
Clonality: Monoclonal
Clone: 238446
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR