Anti-ITGAM Mouse Monoclonal Antibody [clone: ICRF44]

Supplier: R & D Systems
MAB1699
220019-272EA 763.57 USD
220019-272
Anti-ITGAM Mouse Monoclonal Antibody [clone: ICRF44]
Antibodies
Type: Primary
Antigen: ITGAM
Clonality: Monoclonal
Clone: ICRF44
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR