Anti-CD93 Rat Monoclonal Antibody [clone: 223437]

Supplier: R & D Systems
MAB1696
220019-271EA 594.37 USD
220019-271
Anti-CD93 Rat Monoclonal Antibody [clone: 223437]
Antibodies
Type: Primary
Antigen: CD93
Clonality: Monoclonal
Clone: 223437
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR