Anti-CRTAM Mouse Monoclonal Antibody [clone: 210206]

Supplier: R & D Systems
MAB1695
220019-269EA 763.57 USD
220019-269
Anti-CRTAM Mouse Monoclonal Antibody [clone: 210206]
Antibodies
Type: Primary
Antigen: CRTAM
Clonality: Monoclonal
Clone: 210206
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR