Anti-FSTL1 Rat Monoclonal Antibody [clone: 229007]

Supplier: R & D Systems
MAB1694
220019-268EA 772.25 USD
220019-268
Anti-FSTL1 Rat Monoclonal Antibody [clone: 229007]
Antibodies
Type: Primary
Antigen Symbol: FSTL1
Antigen Name: follistatin-like 1
Antigen Synonyms: FRP, FSL1, MIR198
Clonality: Monoclonal
Clone: 229007
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR