Anti-CD58 Mouse Monoclonal Antibody [clone: 248310]

Supplier: R & D Systems
MAB1689
220019-266EA 772.25 USD
220019-266
Anti-CD58 Mouse Monoclonal Antibody [clone: 248310]
Antibodies
Type: Primary
Antigen: CD58
Clonality: Monoclonal
Clone: 248310
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR