Anti-CXCR3 Rat Monoclonal Antibody [clone: 220803]

Supplier: R & D Systems
MAB1685-100
102885-290EA 633.42 USD
102885-290
Anti-CXCR3 Rat Monoclonal Antibody [clone: 220803]
Antibodies
Type: Primary
Antigen: CXCR3
Clonality: Monoclonal
Clone: 220803
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR