Anti-LTB Mouse Monoclonal Antibody [clone: 135105]

Supplier: R & D Systems
MAB1684
220019-263EA 772.25 USD
220019-263
Anti-LTB Mouse Monoclonal Antibody [clone: 135105]
Antibodies
Type: Primary
Antigen: LTB
Clonality: Monoclonal
Clone: 135105
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR