Anti-ICOS Rat Monoclonal Antibody [clone: 670306]

Supplier: R & D Systems
MAB168
102885-288EA 577.02 USD
102885-288
Anti-ICOS Rat Monoclonal Antibody [clone: 670306]
Antibodies
Type: Primary
Antigen: ICOS
Clonality: Monoclonal
Clone: 670306
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR