Anti-FABP3 Rat Monoclonal Antibody [clone: 242305]

Supplier: R & D Systems
MAB1678
220019-261EA 594.37 USD
220019-261
Anti-FABP3 Rat Monoclonal Antibody [clone: 242305]
Antibodies
Type: Primary
Antigen Symbol: FABP3
Antigen Name: fatty acid binding protein 3, muscle and heart
Antigen Synonyms: FABP11, H-FABP, M-FABP, MDGI, O-FABP
Clonality: Monoclonal
Clone: 242305
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR