Anti-GFRA4 Rat Monoclonal Antibody [clone: 171217]

Supplier: R & D Systems
MAB1677
220019-260EA 763.57 USD
220019-260
Anti-GFRA4 Rat Monoclonal Antibody [clone: 171217]
Antibodies
Type: Primary
Antigen Symbol: GFRA4
Antigen Name: glial cell line derived neurotrophic factor family receptor alpha 4
Antigen Synonyms: AW060733, G630015H18Rik
Clonality: Monoclonal
Clone: 171217
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR