Anti-pY Rat Monoclonal Antibody [clone: 216954]

Supplier: R & D Systems
MAB16761
220019-259EA 763.57 USD
220019-259
Anti-pY Rat Monoclonal Antibody [clone: 216954]
Antibodies
Type: Primary
Antigen: pY
Clonality: Monoclonal
Clone: 216954
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR