Anti-PAX7 Mouse Monoclonal Antibody [clone: PAX7]

Supplier: R & D Systems
MAB1675
220019-257EA 655.11 USD
220019-257
Anti-PAX7 Mouse Monoclonal Antibody [clone: PAX7]
Antibodies
Type: Primary
Antigen: PAX7
Clonality: Monoclonal
Clone: PAX7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR