Anti-N-Me-6,7-diOH-TIQ Mouse Monoclonal Antibody [clone: 106814]

Supplier: R & D Systems
MAB167
220019-255EA 772.25 USD
220019-255
Anti-N-Me-6,7-diOH-TIQ Mouse Monoclonal Antibody [clone: 106814]
Antibodies
Type: Primary
Antigen: N-Me-6,7-diOH-TIQ
Clonality: Monoclonal
Clone: 106814
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR