Anti-SERPINA4 Mouse Monoclonal Antibody [clone: 209919]

Supplier: R & D Systems
MAB16691
220019-254EA 772.25 USD
220019-254
Anti-SERPINA4 Mouse Monoclonal Antibody [clone: 209919]
Antibodies
Type: Primary
Antigen: SERPINA4
Clonality: Monoclonal
Clone: 209919
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR