Anti-SERPINA4 Mouse Monoclonal Antibody [clone: 209930]

Supplier: R & D Systems
MAB1669
220019-253EA 763.57 USD
220019-253
Anti-SERPINA4 Mouse Monoclonal Antibody [clone: 209930]
Antibodies
Type: Primary
Antigen: SERPINA4
Clonality: Monoclonal
Clone: 209930
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR