Anti-PAPPA2 Mouse Monoclonal Antibody [clone: 242011]

Supplier: R & D Systems
MAB1668
220019-252EA 772.25 USD
220019-252
Anti-PAPPA2 Mouse Monoclonal Antibody [clone: 242011]
Antibodies
Type: Primary
Antigen: PAPPA2
Clonality: Monoclonal
Clone: 242011
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR