Anti-TPSG1 Rat Monoclonal Antibody [clone: 279005]

Supplier: R & D Systems
MAB1667
220019-251EA 566.89 USD
220019-251
Anti-TPSG1 Rat Monoclonal Antibody [clone: 279005]
Antibodies
Type: Primary
Antigen: TPSG1
Clonality: Monoclonal
Clone: 279005
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR