Anti-STAT2 Mouse Monoclonal Antibody [clone: 545103]

Supplier: R & D Systems
MAB16661
220019-250EA 577.02 USD
220019-250
Anti-STAT2 Mouse Monoclonal Antibody [clone: 545103]
Antibodies
Type: Primary
Antigen: STAT2
Clonality: Monoclonal
Clone: 545103
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR