Anti-MTOR Rat Monoclonal Antibody [clone: 834115]

Supplier: R & D Systems
MAB1665
102885-286EA 577.02 USD
102885-286
Anti-MTOR Rat Monoclonal Antibody [clone: 834115]
Antibodies
Type: Primary
Antigen: MTOR
Clonality: Monoclonal
Clone: 834115
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR