Anti-RTN4R Rat Monoclonal Antibody [clone: 202604]

Supplier: R & D Systems
MAB1659
220019-246EA 763.57 USD
220019-246
Anti-RTN4R Rat Monoclonal Antibody [clone: 202604]
Antibodies
Type: Primary
Antigen: RTN4R
Clonality: Monoclonal
Clone: 202604
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR