Anti-PODXL Mouse Monoclonal Antibody [clone: 222328]

Supplier: R & D Systems
MAB1658
220019-245EA 655.11 USD
220019-245
Anti-PODXL Mouse Monoclonal Antibody [clone: 222328]
Antibodies
Type: Primary
Antigen: PODXL
Clonality: Monoclonal
Clone: 222328
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR