Anti-DUSP3 Mouse Monoclonal Antibody [clone: 237018]

Supplier: R & D Systems
MAB16541
220019-244EA 561.11 USD
220019-244
Anti-DUSP3 Mouse Monoclonal Antibody [clone: 237018]
Antibodies
Type: Primary
Antigen: DUSP3
Clonality: Monoclonal
Clone: 237018
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR