Anti-DUSP3 Mouse Monoclonal Antibody [clone: 237020]

Supplier: R & D Systems
MAB1654
220019-243EA 584.25 USD
220019-243
Anti-DUSP3 Mouse Monoclonal Antibody [clone: 237020]
Antibodies
Type: Primary
Antigen: DUSP3
Clonality: Monoclonal
Clone: 237020
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR