Anti-PPP2CA Rat Monoclonal Antibody [clone: 234532]

Supplier: R & D Systems
MAB16531
220019-242EA 561.11 USD
220019-242
Anti-PPP2CA Rat Monoclonal Antibody [clone: 234532]
Antibodies
Type: Primary
Antigen: PPP2CA
Clonality: Monoclonal
Clone: 234532
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR