Anti-ICOSL Mouse Monoclonal Antibody [clone: 136716]

Supplier: R & D Systems
MAB1651
220019-240EA 763.57 USD
220019-240
Anti-ICOSL Mouse Monoclonal Antibody [clone: 136716]
Antibodies
Type: Primary
Antigen: ICOSL
Clonality: Monoclonal
Clone: 136716
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR