Anti-CDC25A Mouse Monoclonal Antibody [clone: 336445]

Supplier: R & D Systems
MAB1648
220019-238EA 577.02 USD
220019-238
Anti-CDC25A Mouse Monoclonal Antibody [clone: 336445]
Antibodies
Type: Primary
Antigen: CDC25A
Clonality: Monoclonal
Clone: 336445
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR