Anti-KREMEN1 Rat Monoclonal Antibody [clone: 252525]

Supplier: R & D Systems
MAB1647
220019-237EA 763.57 USD
220019-237
Anti-KREMEN1 Rat Monoclonal Antibody [clone: 252525]
Antibodies
Type: Primary
Antigen: KREMEN1
Clonality: Monoclonal
Clone: 252525
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR