Anti-ECE2 Rat Monoclonal Antibody [clone: 233914]

Supplier: R & D Systems
MAB16451
220019-235EA 561.11 USD
220019-235
Anti-ECE2 Rat Monoclonal Antibody [clone: 233914]
Antibodies
Type: Primary
Antigen: ECE2
Clonality: Monoclonal
Clone: 233914
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR