Anti-SLAMF1 Mouse Monoclonal Antibody [clone: 542301]

Supplier: R & D Systems
MAB1642
220019-234EA 633.42 USD
220019-234
Anti-SLAMF1 Mouse Monoclonal Antibody [clone: 542301]
Antibodies
Type: Primary
Antigen: SLAMF1
Clonality: Monoclonal
Clone: 542301
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR