Anti-SLAMF1 Mouse Monoclonal Antibody [clone: 123317]

Supplier: R & D Systems
MAB164
220019-232EA 763.57 USD
220019-232
Anti-SLAMF1 Mouse Monoclonal Antibody [clone: 123317]
Antibodies
Type: Primary
Antigen: SLAMF1
Clonality: Monoclonal
Clone: 123317
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR