Anti-THBS2 Mouse Monoclonal Antibody [clone: 230934]

Supplier: R & D Systems
MAB1635
220019-228EA 772.25 USD
220019-228
Anti-THBS2 Mouse Monoclonal Antibody [clone: 230934]
Antibodies
Type: Primary
Antigen: THBS2
Clonality: Monoclonal
Clone: 230934
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR