Anti-FGF11 Mouse Monoclonal Antibody [clone: 224812]

Supplier: R & D Systems
MAB1634
220019-227EA 772.25 USD
220019-227
Anti-FGF11 Mouse Monoclonal Antibody [clone: 224812]
Antibodies
Type: Primary
Antigen Symbol: FGF11
Antigen Name: fibroblast growth factor 11
Antigen Synonyms: FGF-11, FHF-3, FHF3
Clonality: Monoclonal
Clone: 224812
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR