Anti-OSCAR Rat Monoclonal Antibody [clone: 218311]

Supplier: R & D Systems
MAB1633
220019-226EA 763.57 USD
220019-226
Anti-OSCAR Rat Monoclonal Antibody [clone: 218311]
Antibodies
Type: Primary
Antigen: OSCAR
Clonality: Monoclonal
Clone: 218311
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR