Anti-CARTPT (aa 28-116) Mouse Monoclonal Antibody [clone: 113612]

Supplier: R & D Systems
MAB163
220019-224EA 772.25 USD
220019-224
Anti-CARTPT (aa 28-116) Mouse Monoclonal Antibody [clone: 113612]
Antibodies
Type: Primary
Antigen: CARTPT (aa 28-116)
Clonality: Monoclonal
Clone: 113612
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR