Anti-INHBC Mouse Monoclonal Antibody [clone: 203522]

Supplier: R & D Systems
MAB1629
220019-223EA 772.25 USD
220019-223
Anti-INHBC Mouse Monoclonal Antibody [clone: 203522]
Antibodies
Type: Primary
Antigen: INHBC
Clonality: Monoclonal
Clone: 203522
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR