Anti-WISP1 Mouse Monoclonal Antibody [clone: 213609]

Supplier: R & D Systems
MAB16271
102885-282EA 772.25 USD
102885-282
Anti-WISP1 Mouse Monoclonal Antibody [clone: 213609]
Antibodies
Type: Primary
Antigen: WISP1
Clonality: Monoclonal
Clone: 213609
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR